Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300009757.mm.1 |
NM_001289568.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009757.mm.1 |
NM_001289571.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009757.mm.1 |
NM_028349.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009757.mm.1 |
TC0300001194.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300009759.mm.1 |
NM_001289568.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009759.mm.1 |
NM_028349.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009759.mm.1 |
TC0300001194.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300009760.mm.1 |
NM_001289568.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009760.mm.1 |
TC0300001194.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300009758.mm.1 |
NM_001289568.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009758.mm.1 |
NM_001289571.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009758.mm.1 |
NM_028349.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009758.mm.1 |
TC0300001194.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300009761.mm.1 |
NM_001289568.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009761.mm.1 |
NM_001289571.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009761.mm.1 |
NM_028349.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009761.mm.1 |
TC0300001194.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300009762.mm.1 |
NM_001289568.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009762.mm.1 |
NM_001289571.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009762.mm.1 |
NM_028349.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009762.mm.1 |
TC0300001194.mm.1 |
MTA TC |