Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arrdc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042592.2 | NP_001036057.1 | 2 - 3 | 538 | 92 | |
NM_025549.3 | NP_079825.2 | 2 - 3 | 538 | 92 | ||
GenBank mRNA | AK037198.1 | - | 2 - 3 | 505 | 92 | |
BC017528.1 | - | 2 - 3 | 488 | 4922348 | 92 | |
DQ861996.1 | - | 2 - 3 | 342 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |