Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apba2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291166.1 | NP_001278095.1 | 9 - 10 | 1680 | 68 | |
NM_001291167.1 | NP_001278096.1 | 8 - 9 | 1626 | 68 | ||
NM_007461.2 | NP_031487.1 | 9 - 10 | 1677 | 68 | ||
GenBank mRNA | BC057620.1 | - | 9 - 10 | 1677 | 6402377 | 68 |
BC060269.1 | - | 6 - 7 | 1380 | 6810117 | 68 | |
L34676.1 | - | 6 - 7 | 1139 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |