Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rundc3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198620.1 | NP_941022.1 | 1 - 2 | 464 | 59 | |
GenBank mRNA | BC058808.1 | - | 1 - 2 | 464 | 6491326 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |