Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286481.1 | NP_001273410.1 | 1 - 2 | 220 | 134 | |
NM_001286482.1 | NP_001273411.1 | 1 - 2 | 220 | 134 | ||
GenBank mRNA | BC027104.1 | - | 1 - 2 | 78 | 5359966 | 134 |
BC029667.1 | - | 1 - 2 | 82 | 5362013 | 134 | |
BQ959742.1 | - | 1 - 2 | 220 | 134 | ||
CO424557.1 | - | 1 - 2 | 145 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |