Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec24b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207209.2 | NP_997092.1 | 12 - 13 | 2428 | 155 | |
GenBank mRNA | AK150076.1 | - | 11 - 12 | 1505 | 155 | |
AK152339.1 | - | 10 - 11 | 1400 | 155 | ||
BC046556.1 | - | 12 - 13 | 2260 | 155 | ||
BC046776.1 | - | 12 - 13 | 2428 | 5706007 | 155 | |
BC057355.1 | - | 12 - 13 | 2289 | 6833698 | 155 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |