Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290712.1 | NP_001277641.1 | 4 - 5 | 633 | 89 | |
NM_013686.4 | NP_038714.2 | 5 - 6 | 719 | 89 | ||
GenBank mRNA | AK149611.1 | - | 5 - 6 | 584 | 89 | |
AK149755.1 | - | 5 - 6 | 585 | 89 | ||
AK150666.1 | - | 3 - 4 | 2144 | 89 | ||
AK150744.1 | - | 3 - 4 | 2144 | 89 | ||
AK151068.1 | - | 5 - 6 | 584 | 89 | ||
AK151445.1 | - | 4 - 5 | 523 | 89 | ||
AK151903.1 | - | 3 - 4 | 2144 | 89 | ||
AK152633.1 | - | 3 - 4 | 2144 | 89 | ||
AK152759.1 | - | 5 - 6 | 585 | 89 | ||
AK153037.1 | - | 5 - 6 | 585 | 89 | ||
AK153430.1 | - | 4 - 5 | 523 | 89 | ||
AK165665.1 | - | 5 - 6 | 586 | 89 | ||
AK166828.1 | - | 5 - 6 | 583 | 89 | ||
AK166966.1 | - | 5 - 6 | 607 | 89 | ||
AK167529.1 | - | 5 - 6 | 584 | 89 | ||
BC003809.1 | - | 5 - 6 | 567 | 3599670 | 89 | |
CN725473.1 | - | 4 - 5 | 633 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |