Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Homer2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164086.1 | NP_001157558.1 | 5 - 6 | 659 | 86 | |
NM_001164087.1 | NP_001157559.1 | 5 - 6 | 675 | 86 | ||
NM_011983.2 | NP_036113.1 | 5 - 6 | 692 | 86 | ||
GenBank mRNA | AB017136.1 | - | 5 - 6 | 685 | 86 | |
AF093259.1 | - | 5 - 6 | 564 | 86 | ||
AF093260.1 | - | 5 - 6 | 597 | 86 | ||
AJ012076.1 | - | 5 - 6 | 544 | 86 | ||
AK158673.1 | - | 601 | 86 | |||
BC038314.1 | - | 1 - 2 | 333 | 86 | ||
DV657327.1 | - | 5 - 6 | 597 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |