Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011465.4 | NP_035595.2 | 14 - 15 | 1994 | 156 | |
GenBank mRNA | AF093576.1 | - | 14 - 15 | 1902 | 156 | |
AK142860.1 | - | 14 - 15 | 1969 | 156 | ||
BC150747.1 | - | 14 - 15 | 1859 | 9087658 | 156 | |
U87455.1 | - | 14 - 15 | 1974 | 156 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |