Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011465.4 | NP_035595.2 | 33 - 34 | 4896 | 95 | |
GenBank mRNA | AF093576.1 | - | 33 - 34 | 4804 | 95 | |
AK161449.1 | - | 4 - 5 | 537 | 95 | ||
BC150747.1 | - | 33 - 34 | 4761 | 9087658 | 95 | |
M10276.1 | - | 1 - 2 | 33 | 95 | ||
U87455.1 | - | 33 - 34 | 4876 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |