Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcts1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026902.3 | NP_081178.1 | 3 - 4 | 374 | 120 | |
GenBank mRNA | AK005292.1 | - | 3 - 4 | 375 | 120 | |
AK087975.1 | - | 3 - 4 | 378 | 120 | ||
AK138385.1 | - | 3 - 4 | 393 | 120 | ||
BC010486.1 | - | 3 - 4 | 347 | 3483367 | 120 | |
BC037103.1 | - | 3 - 4 | 363 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |