Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tomm40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109748.1 | NP_001103218.1 | 5 - 6 | 817 | 64 | |
NM_016871.2 | NP_058567.2 | 6 - 7 | 719 | 64 | ||
GenBank mRNA | AF316403.1 | - | 5 - 6 | 652 | 64 | |
AF316404.1 | - | 5 - 6 | 652 | 64 | ||
AK142575.1 | - | 5 - 6 | 413 | 64 | ||
AK150019.1 | - | 5 - 6 | 814 | 64 | ||
AK163759.1 | - | 6 - 7 | 703 | 64 | ||
AK165173.1 | - | 6 - 7 | 714 | 64 | ||
AK170991.1 | - | 6 - 7 | 703 | 64 | ||
AK177130.1 | - | 3 - 4 | 239 | 64 | ||
AK187895.1 | - | 3 - 4 | 238 | 64 | ||
AK205085.1 | - | 2 - 3 | 149 | 64 | ||
BC100452.1 | - | 6 - 7 | 707 | 6532792 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |