Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emcn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163522.1 | NP_001156994.1 | 6 - 7 | 674 | 65 | |
NM_016885.2 | NP_058581.2 | 5 - 6 | 635 | 65 | ||
GenBank mRNA | AB034693.1 | - | 6 - 7 | 627 | 65 | |
AF060883.1 | - | 5 - 6 | 570 | 65 | ||
AK002616.1 | - | 5 - 6 | 617 | 65 | ||
AK029348.1 | - | 641 | 65 | |||
AK145825.1 | - | 5 - 6 | 549 | 65 | ||
BC003706.1 | - | 6 - 7 | 635 | 3485788 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |