Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc16b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024645.1 | NP_001019816.1 | 12 - 13 | 1101 | 70 | |
GenBank mRNA | AK131164.1 | - | 12 - 13 | 1044 | 70 | |
AK148355.1 | - | 12 - 13 | 1124 | 70 | ||
AK158466.1 | - | 13 - 14 | 1347 | 70 | ||
BC082805.1 | - | 12 - 13 | 1021 | 70 | ||
BC092223.1 | - | 12 - 13 | 1019 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |