Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: R3hcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146012.2 | NP_001139484.2 | 6 - 7 | 1336 | 132 | |
NM_001301651.1 | NP_001288580.1 | 6 - 7 | 1288 | 132 | ||
NM_001301652.1 | NP_001288581.1 | 5 - 6 | 1569 | 132 | ||
GenBank mRNA | AK006177.1 | - | 6 - 7 | 1288 | 132 | |
AK032854.1 | - | 6 - 7 | 1306 | 132 | ||
AK156843.1 | - | 6 - 7 | 1306 | 132 | ||
AK208982.1 | - | 2 - 3 | 166 | 132 | ||
BC052536.1 | - | 5 - 6 | 1485 | 132 | ||
BC059860.1 | - | 5 - 6 | 1569 | 132 | ||
BQ747890.1 | - | 2 - 3 | 200 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |