Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttll13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177765.3 | NP_808433.2 | 6 - 7 | 1004 | 67 | |
GenBank mRNA | AK029565.1 | - | 6 - 7 | 1003 | 67 | |
AK161519.1 | - | 6 - 7 | 827 | 67 | ||
AM690757.1 | - | 5 - 6 | 705 | 67 | ||
BC145749.1 | - | 5 - 6 | 612 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |