Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnni3k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177066.5 | NP_796040.3 | 5 - 6 | 550 | 76 | |
GenBank mRNA | AY526095.1 | - | 5 - 6 | 550 | 76 | |
AY526096.1 | - | 5 - 6 | 550 | 76 | ||
BC139368.1 | - | 5 - 6 | 483 | 8862390 | 76 | |
BC139394.1 | - | 5 - 6 | 483 | 8862416 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |