Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159516.1 | NP_001152988.1 | 6 - 7 | 1424 | 93 | |
GenBank mRNA | AF090403.1 | - | 7 - 8 | 1584 | 93 | |
AK132303.1 | - | 6 - 7 | 1425 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mus musculus 1700 Inventoried Probe spans exons |