Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080443.2 | NP_536691.2 | 4 - 5 | 930 | 69 | |
GenBank mRNA | BC085155.1 | - | 4 - 5 | 910 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |