Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023233.3 | NP_075722.1 | 1 - 2 | 61 | 65 | |
GenBank mRNA | BM933374.1 | - | 1 - 2 | 61 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |