Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm35066 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK035404.1 | - | 2 - 3 | 367 | 90 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | chloride channel calcium activated 4A | Clca4a |
9130020L07Rik, AI504701, Clca4, Clca6 |
-5.4kb | Anti-sense | Chr. 3 - 144952486 - 144975045 on NCBI Build GRCm38 |
Downstream | chloride channel calcium activated 1 | Clca1 |
Clca2, Clca3, gob-5, gob5 |
23.3kb | Anti-sense | Chr. 3 - 145004537 - 145032776 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Probe spans exons |