Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700029420.mm.1 |
NM_027413.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029420.mm.1 |
NM_027413.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029420.mm.1 |
NM_175124.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029420.mm.1 |
NM_175124.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029420.mm.1 |
NR_028143.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029420.mm.1 |
NR_028143.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029420.mm.1 |
NR_028144.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029420.mm.1 |
NR_028144.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029420.mm.1 |
TC0700003448.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029422.mm.1 |
NM_027413.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029422.mm.1 |
NM_027413.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029422.mm.1 |
NM_175124.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029422.mm.1 |
NM_175124.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029422.mm.1 |
NR_028143.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029422.mm.1 |
NR_028143.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029422.mm.1 |
NR_028144.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029422.mm.1 |
NR_028144.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029422.mm.1 |
TC0700003448.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029424.mm.1 |
NM_027413.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029424.mm.1 |
NM_027413.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029424.mm.1 |
NM_175124.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029424.mm.1 |
NM_175124.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029424.mm.1 |
NR_028143.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029424.mm.1 |
NR_028143.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029424.mm.1 |
NR_028144.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029424.mm.1 |
NR_028144.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029424.mm.1 |
TC0700003448.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029423.mm.1 |
NM_027413.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029423.mm.1 |
NM_027413.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029423.mm.1 |
NM_175124.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029423.mm.1 |
NM_175124.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029423.mm.1 |
NR_028143.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029423.mm.1 |
NR_028143.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029423.mm.1 |
NR_028144.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029423.mm.1 |
NR_028144.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029423.mm.1 |
TC0700003448.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029425.mm.1 |
NM_027413.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029425.mm.1 |
NM_027413.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029425.mm.1 |
NM_175124.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029425.mm.1 |
NM_175124.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029425.mm.1 |
NR_028143.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029425.mm.1 |
NR_028143.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029425.mm.1 |
NR_028144.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029425.mm.1 |
NR_028144.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029425.mm.1 |
TC0700003448.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700029426.mm.1 |
NM_027413.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029426.mm.1 |
NM_027413.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029426.mm.1 |
NM_175124.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029426.mm.1 |
NM_175124.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029426.mm.1 |
NR_028143.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029426.mm.1 |
NR_028143.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029426.mm.1 |
NR_028144.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029426.mm.1 |
NR_028144.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700029426.mm.1 |
TC0700003448.mm.1 |
MTA TC |