Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ska3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198605.3 | NP_941007.1 | 1 - 2 | 155 | 142 | |
GenBank mRNA | AK089223.1 | - | 1 - 2 | 156 | 142 | |
AK158142.1 | - | 1 - 2 | 177 | 142 | ||
BC117500.1 | - | 1 - 2 | 147 | 40105921 | 142 | |
BC117501.1 | - | 1 - 2 | 147 | 40105920 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |