Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136074.2 | NP_001129546.1 | 2 - 3 | 159 | 88 | |
NM_001271916.1 | NP_001258845.1 | 2 - 3 | 179 | 88 | ||
NM_008736.3 | NP_032762.1 | 1 - 2 | 158 | 88 | ||
GenBank mRNA | AK044275.1 | - | 1 - 2 | 123 | 88 | |
AK044316.1 | - | 2 - 3 | 142 | 88 | ||
BC031440.1 | - | 2 - 3 | 143 | 4527590 | 88 | |
L14935.1 | - | 1 - 2 | 97 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |