Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012012.4 | NP_036142.2 | 7 - 8 | 908 | 84 | |
GenBank mRNA | AJ238213.1 | - | 5 - 6 | 763 | 84 | |
AK028728.1 | - | 7 - 8 | 889 | 84 | ||
AK166425.1 | - | 7 - 8 | 884 | 84 | ||
BC006671.1 | - | 7 - 8 | 872 | 3499406 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |