Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnaseh2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026001.2 | NP_080277.1 | 8 - 9 | 702 | 94 | |
GenBank mRNA | AK011914.1 | - | 8 - 9 | 741 | 94 | |
AK082752.1 | - | 8 - 9 | 703 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |