Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026041.2 | NP_080317.3 | 3 - 4 | 556 | 91 | |
GenBank mRNA | AK007616.1 | - | 3 - 4 | 510 | 91 | |
AK010658.1 | - | 3 - 4 | 524 | 91 | ||
AK013217.1 | - | 3 - 4 | 524 | 91 | ||
AK087997.1 | - | 3 - 4 | 526 | 91 | ||
AK169088.1 | - | 3 - 4 | 557 | 91 | ||
BC006787.1 | - | 3 - 4 | 522 | 3589453 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |