Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020010.2 | NP_064394.2 | 1 - 2 | 563 | 82 | |
GenBank mRNA | AF166266.1 | - | 1 - 2 | 315 | 82 | |
AK028355.1 | - | 1 - 2 | 256 | 82 | ||
AK028815.1 | - | 1 - 2 | 320 | 82 | ||
AK031059.1 | - | 1 - 2 | 296 | 82 | ||
AK075764.1 | - | 1 - 2 | 296 | 82 | ||
AK076983.1 | - | 1 - 2 | 296 | 82 | ||
BC031813.1 | - | 1 - 2 | 290 | 4193738 | 82 | |
CB524876.1 | - | 1 - 2 | 563 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |