Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crtc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173863.2 | NP_776288.2 | 12 - 13 | 1610 | 62 | |
GenBank mRNA | AK144840.1 | - | 12 - 13 | 1612 | 62 | |
BC011210.1 | - | 12 - 13 | 1449 | 4195538 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |