Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016676.5 | NP_057885.1 | 2 - 3 | 744 | 139 | |
GenBank mRNA | AB232604.1 | - | 2 - 3 | 193 | 139 | |
AF035646.1 | - | 2 - 3 | 434 | 139 | ||
AK008725.1 | - | 2 - 3 | 734 | 139 | ||
AK028320.1 | - | 2 - 3 | 715 | 139 | ||
AK087964.1 | - | 2 - 3 | 706 | 139 | ||
AK134988.1 | - | 2 - 3 | 664 | 139 | ||
AK146357.1 | - | 2 - 3 | 745 | 139 | ||
AK150319.1 | - | 2 - 3 | 704 | 139 | ||
AK150384.1 | - | 2 - 3 | 398 | 139 | ||
AK150791.1 | - | 2 - 3 | 694 | 139 | ||
AK151473.1 | - | 2 - 3 | 398 | 139 | ||
AK153315.1 | - | 2 - 3 | 704 | 139 | ||
AK153325.1 | - | 2 - 3 | 694 | 139 | ||
AK159383.1 | - | 2 - 3 | 709 | 139 | ||
AK159845.1 | - | 2 - 3 | 709 | 139 | ||
AK159988.1 | - | 2 - 3 | 703 | 139 | ||
AK160960.1 | - | 2 - 3 | 699 | 139 | ||
AK166010.1 | - | 2 - 3 | 715 | 139 | ||
AK168320.1 | - | 2 - 3 | 704 | 139 | ||
AK195457.1 | - | 2 - 3 | 180 | 139 | ||
BC052735.1 | - | 694 | 139 | |||
BC056374.1 | - | 2 - 3 | 695 | 6848840 | 139 | |
CT010394.1 | - | 2 - 3 | 193 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |