Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033314.3 | NP_001028486.1 | 6 - 7 | 1039 | 78 | |
GenBank mRNA | AK082980.1 | - | 6 - 7 | 1004 | 78 | |
AK146297.1 | - | 6 - 7 | 1040 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |