Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153422.2 | NP_700471.2 | 10 - 11 | 1627 | 59 | |
GenBank mRNA | AF541937.1 | - | 10 - 11 | 1627 | 59 | |
BC119137.1 | - | 10 - 11 | 1590 | 8733886 | 59 | |
BC120486.1 | - | 10 - 11 | 1590 | 40129409 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |