Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030112.2 | NP_084388.2 | 15 - 16 | 1837 | 60 | |
GenBank mRNA | AK135633.1 | - | 9 - 10 | 905 | 60 | |
BC117848.1 | - | 15 - 16 | 1689 | 40093717 | 60 | |
BC117849.1 | - | 15 - 16 | 1689 | 40093724 | 60 | |
CN532066.1 | - | 5 - 6 | 353 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |