Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rreb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039188.1 | NP_001034277.1 | 7 - 8 | 1159 | 63 | |
NM_001177868.1 | NP_001171339.1 | 7 - 8 | 884 | 63 | ||
NM_001177869.1 | NP_001171340.1 | 6 - 7 | 820 | 63 | ||
NM_026830.2 | NP_081106.1 | 7 - 8 | 1127 | 63 | ||
NR_033218.1 | 7 - 8 | 2281 | 63 | |||
NR_033615.1 | 7 - 8 | 2098 | 63 | |||
GenBank mRNA | AK047047.1 | - | 7 - 8 | 2078 | 63 | |
AK047251.1 | - | 7 - 8 | 2080 | 63 | ||
AK147260.1 | - | 7 - 8 | 2282 | 63 | ||
AK147653.1 | - | 7 - 8 | 2282 | 63 | ||
AK154980.1 | - | 7 - 8 | 1127 | 63 | ||
AK170652.1 | - | 7 - 8 | 1159 | 63 | ||
AK171375.1 | - | 7 - 8 | 864 | 63 | ||
AY946044.1 | - | 4 - 5 | 511 | 63 | ||
BC080680.1 | - | 7 - 8 | 973 | 6330949 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |