Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tax1bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025816.3 | NP_080092.2 | 16 - 17 | 2291 | 68 | |
GenBank mRNA | AK004574.1 | - | 16 - 17 | 2273 | 68 | |
AK146076.1 | - | 16 - 17 | 2271 | 68 | ||
BC014798.1 | - | 16 - 17 | 2212 | 3962810 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |