Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cts7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019539.3 | NP_062412.1 | 4 - 5 | 611 | 92 | |
GenBank mRNA | AF250837.1 | - | 3 - 4 | 398 | 92 | |
AY014779.1 | - | 4 - 5 | 471 | 92 | ||
BC050198.1 | - | 3 - 4 | 707 | 92 | ||
BC064740.1 | - | 4 - 5 | 611 | 30018572 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |