Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc34a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011392.2 | NP_035522.2 | 5 - 6 | 645 | 80 | |
GenBank mRNA | AB163921.1 | - | 650 | 80 | ||
AB163922.1 | - | 5 - 6 | 589 | 80 | ||
AK018740.2 | - | 5 - 6 | 600 | 80 | ||
BC020529.1 | - | 5 - 6 | 588 | 4223484 | 80 | |
L33878.1 | - | 4 - 5 | 572 | 80 | ||
U22465.1 | - | 5 - 6 | 589 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |