Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Urm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026615.4 | NP_080891.1 | 2 - 3 | 114 | 93 | |
GenBank mRNA | AK019352.1 | - | 2 - 3 | 120 | 93 | |
AK049227.1 | - | 2 - 3 | 93 | |||
AK049849.1 | - | 2 - 3 | 115 | 93 | ||
AK210360.1 | - | 2 - 3 | 98 | 93 | ||
BC026994.1 | - | 2 - 3 | 106 | 5368627 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |