Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Degs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027299.5 | NP_081575.2 | 2 - 3 | 888 | 72 | |
GenBank mRNA | AF466377.1 | - | 2 - 3 | 876 | 72 | |
AK008666.1 | - | 2 - 3 | 889 | 72 | ||
BC016427.1 | - | 2 - 3 | 870 | 4218832 | 72 | |
BC035454.1 | - | 2 - 3 | 870 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |