Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sel1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033296.2 | NP_001028468.1 | 14 - 15 | 1332 | 96 | |
GenBank mRNA | AK016809.1 | - | 14 - 15 | 1278 | 96 | |
AK132650.1 | - | 14 - 15 | 1333 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |