Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252454.1 | NP_001239383.1 | 3 - 4 | 745 | 100 | |
NM_010147.4 | NP_034277.1 | 4 - 5 | 916 | 100 | ||
GenBank mRNA | AK155033.1 | - | 4 - 5 | 911 | 100 | |
BC046962.1 | - | 2 - 3 | 551 | 6391968 | 100 | |
BC067206.1 | - | 3 - 4 | 687 | 6514327 | 100 | |
BC099682.1 | - | 4 - 5 | 846 | 6529672 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |