Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbs5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028284.2 | NP_082560.1 | 5 - 6 | 416 | 102 | |
GenBank mRNA | AK012283.1 | - | 5 - 6 | 416 | 102 | |
BC061031.1 | - | 5 - 6 | 417 | 6771365 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |