Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008625.2 | NP_032651.2 | 5 - 6 | 1019 | 73 | |
GenBank mRNA | BC141338.1 | - | 5 - 6 | 971 | 9055950 | 73 |
Z11974.1 | - | 5 - 6 | 1019 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |