Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rassf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175445.4 | NP_780654.1 | 1 - 2 | 241 | 63 | |
GenBank mRNA | AK028384.1 | - | 1 - 2 | 145 | 63 | |
BC057402.1 | - | 1 - 2 | 63 | 5698333 | 63 | |
BQ770520.1 | - | 1 - 2 | 241 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |