Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haus2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290807.1 | NP_001277736.1 | 3 - 4 | 219 | 110 | |
NM_025475.3 | NP_079751.1 | 3 - 4 | 319 | 110 | ||
GenBank mRNA | AK007106.1 | - | 3 - 4 | 221 | 110 | |
AK010445.1 | - | 3 - 4 | 266 | 110 | ||
AK012086.1 | - | 3 - 4 | 318 | 110 | ||
AK012960.1 | - | 3 - 4 | 266 | 110 | ||
AK132300.1 | - | 3 - 4 | 275 | 110 | ||
BC086677.1 | - | 3 - 4 | 257 | 30660268 | 110 | |
BY711279.1 | - | 3 - 4 | 321 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |