Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200030490.mm.1 |
NM_001110204.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200030490.mm.1 |
NM_001110204.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200030490.mm.1 |
NM_001110205.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200030490.mm.1 |
NM_001110205.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200030490.mm.1 |
NM_007394.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200030490.mm.1 |
NM_007394.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200030490.mm.1 |
TC0200003579.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200030483.mm.1 |
NM_001110204.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200030483.mm.1 |
NM_001110204.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200030483.mm.1 |
NM_001110205.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200030483.mm.1 |
NM_001110205.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200030483.mm.1 |
NM_007394.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200030483.mm.1 |
NM_007394.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200030483.mm.1 |
TC0200003579.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200030488.mm.1 |
NM_001110204.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200030488.mm.1 |
NM_001110204.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200030488.mm.1 |
NM_001110205.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200030488.mm.1 |
NM_001110205.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200030488.mm.1 |
NM_007394.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200030488.mm.1 |
NM_007394.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200030488.mm.1 |
TC0200003579.mm.1 |
MTA TC |