Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Orc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011958.3 | NP_036088.3 | 9 - 10 | 918 | 145 | |
GenBank mRNA | AJ003140.1 | - | 9 - 10 | 879 | 145 | |
AK010258.1 | - | 9 - 10 | 902 | 145 | ||
AK076324.1 | - | 9 - 10 | 919 | 145 | ||
AK145454.1 | - | 9 - 10 | 929 | 146 | ||
AK169291.1 | - | 9 - 10 | 902 | 145 | ||
BC015072.1 | - | 9 - 10 | 890 | 4212266 | 145 | |
BC019748.1 | - | 9 - 10 | 860 | 5007346 | 145 | |
Y16386.1 | - | 8 - 9 | 750 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |