Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acvr2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007396.4 | NP_031422.3 | 1 - 2 | 718 | 64 | |
GenBank mRNA | AK041246.1 | - | 1 - 2 | 671 | 64 | |
AK143186.1 | - | 1 - 2 | 626 | 64 | ||
BC138823.1 | - | 1 - 2 | 379 | 8861845 | 64 | |
M65287.1 | - | 1 - 2 | 125 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |