Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tor1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133673.3 | NP_598434.2 | 3 - 4 | 665 | 67 | |
GenBank mRNA | AJ297743.1 | - | 3 - 4 | 695 | 67 | |
AK017290.1 | - | 3 - 4 | 683 | 67 | ||
AK028859.1 | - | 3 - 4 | 480 | 67 | ||
AK054404.1 | - | 3 - 4 | 696 | 67 | ||
AK149754.1 | - | 3 - 4 | 686 | 67 | ||
AK159843.1 | - | 3 - 4 | 664 | 67 | ||
AK169692.1 | - | 2 - 3 | 2448 | 67 | ||
AK171756.1 | - | 1 - 2 | 1691 | 67 | ||
BC018456.1 | - | 3 - 4 | 664 | 3582246 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |