Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efcab11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030172.2 | NP_084448.1 | 1 - 2 | 154 | 64 | |
GenBank mRNA | AK011507.1 | - | 1 - 2 | 154 | 64 | |
AK011836.1 | - | 1 - 2 | 136 | 64 | ||
AK035051.1 | - | 1 - 2 | 134 | 64 | ||
AK132473.1 | - | 1 - 2 | 216 | 64 | ||
BC049109.1 | - | 1 - 2 | 116 | 6414718 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |